D-(+)-Trehalose, anhydrous
D-(+)-Trehalose, anhydrous
D-(+)-Trehalose, anhydrous
D-(+)-Trehalose, anhydrous
Thermo Scientific Chemicals
D-(+)-Trehalose, anhydrous
CAS: 99-20-7 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFJ66006.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-(+)-Trehalose
Name Noteanhydrous
CAS99-20-7
ColorWhite
Recommended StorageAmbient temperatures; Store under Argon
View more
Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Anthony Kennedy, Edward Ross Pennington. Conservation of chemically degraded waterlogged wood with sugars. Studies in Conservation. 2014, 59, (3), 194-201.
  2. Crowe, L.M.; Reid, D.S.; Crowe, J.H. Is trehalose special for preserving dry biomaterials. Biophys J. 1996, 71, (4), 2087-2093.