Biebrich Scarlet sodium salt
Biebrich Scarlet sodium salt
Biebrich Scarlet sodium salt
Biebrich Scarlet sodium salt
Thermo Scientific Chemicals
Biebrich Scarlet sodium salt
CAS: 4196-99-0 | C22H14N4Na2O7S2 | 556.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFJ65603.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBiebrich Scarlet sodium salt
Melting Point181°C to 188°C
CAS4196-99-0
Recommended StorageAmbient temperatures
EINECS Number224-084-5