Glycine, Electrophoresis Grade, 99+%
Glycine, Electrophoresis Grade, 99+%
Glycine, Electrophoresis Grade, 99+%
Glycine, Electrophoresis Grade, 99+%
Thermo Scientific Chemicals
Glycine, Electrophoresis Grade, 99+%
CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALFJ64365.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycine
Melting Point∽245°C (decomposition)
CAS56-40-6
pH5.97
ColorWhite
View more
Glycine is used as a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin?s transfer buffer for Western blots. Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycine is used as a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin′s transfer buffer for Western blots. Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
Solubility
Freely soluble in water
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Tapio Haaparanta.; Jouni Uitto.; Erkki Ruoslahti.; Eva Engvall. A determination of submicromolar concentrations of glycine in periaqueductal gray matter microdialyzates using capillary zone electrophoresis with contactless conductivity detection. Electrophoresis. 2009, 30, (19), 3436-3441.