Wortmannin, Penicillium funiculosum, 99+%
Wortmannin, Penicillium funiculosum, 99+%
Wortmannin, Penicillium funiculosum, 99+%
Wortmannin, Penicillium funiculosum, 99+%
Thermo Scientific Chemicals
Wortmannin, Penicillium funiculosum, 99+%
Inhibitor of phosphatidylinositol 3-kinase | CAS: 19545-26-7 | C23H24O8, C23H24O8 | 428.44 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63983.MA10 mg
Catalog number ALFJ63983.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Specifications
Quantity10 mg
DescriptionWhite to Off-white
For Use With (Application)Inhibitor of phosphatidylinositol 3-kinase
Recommended StorageStore at -20°C
Assay≥99%
View more
Wortmannin is used as inhibitor of phosphatidylinositol 3-kinase, used cell biology reagent that has been used previously in research to inhibit DNA repair, receptor-mediated endocytosis and cell proliferation. It inhibits fMLP induced PIP3 and superoxide anion production in guinea pig neutrophils. It blocks insulin metabolic effects in rat adipocytes without affecting the insulin receptor tyrosine kinase activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C