Tulobuterol
Tulobuterol
Actual product may vary
Tulobuterol
beta-2-adrenergic receptor agonist
Have Questions?
Catalog NumberQuantity
ALFJ63975.031 g
Catalog number ALFJ63975.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity1 g
DescriptionWhite to Off-white
For Use With (Application)beta-2-adrenergic receptor agonist
CAS41570-61-0
Molecular FormulaC12H18ClNO
Formula Weight227.73g/mol
Melting Point159°C to 163°C
Product TypeSignal Transduction Reagent
FormCrystalline Powder
Unit SizeEA
Contents & Storage
-30°C to -10°C