Ticlopidine Hydrochloride
Ticlopidine Hydrochloride
Ticlopidine Hydrochloride
Ticlopidine Hydrochloride
Thermo Scientific Chemicals
Ticlopidine Hydrochloride
Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines | CAS: 53885-35-1 | C14H14ClNS·HCl
Have Questions?
Catalog NumberQuantity
ALFJ63971.065 g
Catalog number ALFJ63971.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Quantity5 g
DescriptionWhite
For Use With (Application)Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines
Recommended StorageAmbient temperatures
CAS53885-35-1
View more
Ticlopidine hydrochloride is a selective P2Y12 receptor antagonist. It inhibits ADP-induced platelet aggregation and displays antithrombotic activity following oral administration.