Sodium citrate, 0.5M buffer soln., pH 6.5
Sodium citrate, 0.5M buffer soln., pH 6.5
Thermo Scientific Chemicals
Sodium citrate, 0.5M buffer soln., pH 6.5
Have Questions?
Catalog NumberQuantity
ALFJ63888.AK250 mL
Catalog number ALFJ63888.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium citrate buffer soln.
ColorColorless
Recommended StorageKeep cold
Concentration0.5 M
TSCAYes
View more
Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 
Solubility
Soluble in water.
Notes
20X SSC Solution (RNase-free) should be stored at room temperature. This reagent should remain stable for at least 1 year in its unopened container.
RUO – Research Use Only
General References:
  1. CJ Quaife.; SD Findley.; JC Erickson.; GJ Froelick. Induction of a new metallothionein isoform (MT-IV) occurs during differentiation of stratified squamous epithelia. Biochemistry. 1994, 33,(23), 7250-7259.
  2. M Cappello.; S Keshav.; C Prince.; DP Jewell.; S Gordon. Detection of mRNAs for macrophage products in inflammatory bowel disease by in situ hybridization. Gut . 1992, 33,(9), 1214-1219.