L-Cystine dihydrochloride, 98%
L-Cystine dihydrochloride, 98%
L-Cystine dihydrochloride, 98%
L-Cystine dihydrochloride, 98%
Thermo Scientific Chemicals
L-Cystine dihydrochloride, 98%
CAS: 30925-07-6 | C6H14Cl2N2O4S2 | 313.208 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63717.A11 kg
Catalog number ALFJ63717.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Cystine dihydrochloride
Melting Point228°C to 232°C
CAS30925-07-6
ColorWhite to Yellow
Recommended StorageAmbient temperatures
View more
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.
Solubility
Freely soluble in Dilute hydrochloric acid.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Yukio Kurita.; Walter Gordy. Electron Spin Resonance in a Gamma-Irradiated Single Crystal of L-Cystine Dihydrochloride.J. Chem. Phys. 1961, 34 (1),282 .
  2. Kazuyuki Akasaka.; Shun-ichi Ohnishi.; Tokuo Suita. ;Isamu Nitta. ESR Study of a Single Crystal of L-Cystine Dihydrochloride Irradiated at Low Temperature.J. Chem. Phys. 1964, 40, (40),3110 .