2,9-Dimethyl-1,10-phenanthroline, 98%
2,9-Dimethyl-1,10-phenanthroline, 98%
2,9-Dimethyl-1,10-phenanthroline, 98%
2,9-Dimethyl-1,10-phenanthroline, 98%
Thermo Scientific Chemicals
2,9-Dimethyl-1,10-phenanthroline, 98%
Reagent for the colorimetric determination of copper | CAS: 484-11-7 | C14H12N2 | 208.264 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFJ63698.14
Price (MYR)
1,709.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,709.00
EA
Add to cart
Specifications
Chemical Name or Material2,9-Dimethyl-1,10-phenanthroline
CAS484-11-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.
Solubility
Slightly soluble in water and light petroleum. Soluble in methanol, ethanol, acetone, ether and benzene.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Ashbrook, L. N.; Elliott, C. M. Dye-Sensitized Solar Cell Studies of a Donor-Appended Bis(2,9-dimethyl-1,10-phenanthroline) Cu(I) Dye Paired with a Cobalt-Based Mediator. J. Phys. Chem. C 2013, 117 (8), 3853-3864.
  2. Xiao, C-L.; Wang, C-Z.; Yuan, L-Y.; Li, B.; He, H.; Wang, S.; Zhao, Y-L.; Chai, Z-F.; Shi, W-Q. Excellent Selectivity for Actinides with a Tetradentate 2,9-Diamide-1,10-Phenanthroline Ligand in Highly Acidic Solution: A Hard-Soft Donor Combined Strategy. Inorg. Chem. 2014, 53 (3), 1712-1720.
  3. Ashbrook, L. N.; Elliott, C. M. Dye-Sensitized Solar Cell Studies of a Donor-Appended Bis(2,9-dimethyl-1,10-phenanthroline) Cu(I) Dye Paired with a Cobalt-Based Mediator. J. Phys. Chem. C. 2013, 117 (8), 3853-3864.