Citrate-buffered saline (20X)
Citrate-buffered saline (20X)
Actual product may vary
Citrate-buffered saline (20X)
Citrate-buffered saline (20X)
Citrate-buffered saline (20X)
Have Questions?
Catalog NumberQuantity
ALFJ63666.K21 L
Catalog number ALFJ63666.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Concentration20X
Molecular Weight (g/mol)294.10
Quantity1 L
CAS6132-04-3
SynonymSSC buffer, 20X; Saline-sodium citrate buffer, 20X
FormLiquid
Unit SizeEA
Contents & Storage
Ambient Temperatures