Vinblastine Sulfate, 98%
Vinblastine Sulfate, 98%
Vinblastine Sulfate, 98%
Vinblastine Sulfate, 98%
Thermo Scientific Chemicals
Vinblastine Sulfate, 98%
Anticancer agent; microtubule disrupter; Induces apoptosis | CAS: 143-67-9 | C46H60N4O13S, C46H60N4O13S | 909.06 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63598.MA10 mg
Catalog number ALFJ63598.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Specifications
Quantity10 mg
DescriptionWhite
For Use With (Application)Anticancer agent; microtubule disrupter; Induces apoptosis
Recommended StorageKeep cold
Assay98%
View more
Anticancer agent; microtubule disrupter; Induces apoptosis.Vinblastine sulfate is therapeutically used as an antineoplastic. It is also used in the treatment of Hodgkin disease, choriocarcinoma, acute and chronic leukemias.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C