Tween 20 washing buffer, 0.5% in PBS (10X)
Tween 20 washing buffer, 0.5% in PBS (10X)
Thermo Scientific Chemicals
Tween 20 washing buffer, 0.5% in PBS (10X)
Have Questions?
Catalog NumberQuantity
ALFJ63596.AP500 mL
Catalog number ALFJ63596.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Product TypeBuffer
Quantity500 mL
FormLiquid
Concentration0.5% in PBS (10X)
Unit SizeEA
Tween 20 washing buffer, 0.5% in PBS (10X) is used as a wash solution for western blot membranes and microtiter plate wells in ELISA assays. It acts as a pH stabilizer in optimal formulation, salts and detergents designed to effectively remove excess material from membranes. It finds application as laboratory detergents as well as in biomedical experiments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient temperatures