D-erythro-Sphingosine, 99+%
D-erythro-Sphingosine, 99+%
D-erythro-Sphingosine, 99+%
D-erythro-Sphingosine, 99+%
Thermo Scientific Chemicals
D-erythro-Sphingosine, 99+%
CAS: 123-78-4 | C18H37NO2 | 299.499 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63584.MB25 mg
Catalog number ALFJ63584.MB
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialD-erythro-Sphingosine
CAS123-78-4
ColorWhite
Recommended StorageStore at -20°C
EINECS Number204-651-3
View more
D-erythro-Sphingosine is used to inhibits protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-erythro-Sphingosine is used to inhibits protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.
Solubility
Soluble in ethanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Hao Yang.; Lanny S. Liebeskind. A Concise and Scalable Synthesis of High Enantiopurity (-)-d-erythro-Sphingosine Using Peptidyl Thiol Ester-Boronic Acid Cross-Coupling.Org. Lett.2007, (16), 2993-2995.
  2. Peter Somfai.; Roger Olsson. An alternative synthesis of d-erythro-sphingosine and l-lyxo-phytosphingosineg.Tetrahedron Letters.1993, (30), 6645-6650.