trans-Zeatin-riboside
trans-Zeatin-riboside
trans-Zeatin-riboside
trans-Zeatin-riboside
Thermo Scientific Chemicals
trans-Zeatin-riboside
CAS: 6025-53-2 | C15H21N5O5 | 351.363 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63382.MF50 mg
Catalog number ALFJ63382.MF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-Zeatin-riboside
CAS6025-53-2
ColorWhite
Recommended StorageKeep cold
TSCANo
View more
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.
Solubility
Soluble in water, ethanol, and methanol.
Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Carl E. Hansen.; Herman Wenzler.; Frederick Meins Jr.. Concentration Gradients of trans-Zeatin Riboside and trans-Zeatin in the Maize Stem.Plant Physiology.1984, 75, (4), 959-963.
  2. Armstrong, D.J.; Burrows, W.J.; Evans, P.K.; Skoog, F. Isolation of cytokinins from tRNA.Biochemical and Biophysical Research Communications.1969, 37, (3), 451-456.