L-(-)-Malic acid, 99%
L-(-)-Malic acid, 99%
L-(-)-Malic acid, 99%
L-(-)-Malic acid, 99%
Thermo Scientific Chemicals
L-(-)-Malic acid, 99%
CAS: 97-67-6 | C4H6O5 | 134.087 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63221.22100 g
Catalog number ALFJ63221.22
Price (MYR)
236.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
236.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-(-)-Malic acid
CAS97-67-6
Health Hazard 1H315-H318-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
View more
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.
Solubility
Soluble in water(363g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Isao Takata; Kozo Yamamoto; Tetsuya Tosa; Ichiro Chibata. Immobilization of Brevibacterium flavum with carrageenan and its application for continuous production of l-malic acid.Enzyme and Microbial Technology.1980, 2, 30-36.
  2. F. A. T. Anet. The Configuration of Deuterio-l-malic acid Produced Enzymatically. Synthesis of Threo-3-deuterio-dl-malic acid. J. Am. Chem. Soc.1960, 82, (4), 994-995.