Polymixin B sulfate
Polymixin B sulfate
Polymixin B sulfate
Thermo Scientific Chemicals
Polymixin B sulfate
Polymixin B sulfate, CAS # 1405-20-5, is a short polypeptide with antibacterial activity against gram-negative bacteria. | CAS: 1405-20-5 | C48H84N16O17S | 1301.571 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63074.031g
Catalog number ALFJ63074.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1g
Request bulk or custom format
Specifications
CAS1405-20-5
FormatPowder
Molecular FormulaC48H84N16O17S
MDL NumberMFCD00131991
Molecular Weight (g/mol)1301.571
View more
Antibiotic for gram-negative bacteria Polymyxin B Sulfate is an antibiotic mixture active against gram (-) organisms. It is also reported to inhibit Ca2+ dependent PKC (protein kinase C).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • Polymixin B sulfate is a mixture of B1 and B2 polypeptides obtained from different strains of Bacillus polymyxa, with antibacterial activity against gram-negative bacteria. It does not exhibit activity against fungi and gram-positive organisms
  • This compound inhibits Ca2+ dependent PKC (protein kinase C)
  • It can bind lipopolysaccharide (LPS) of the outer membrane of gram-negative bacteria and disrupts the cytoplasmic membrane by inducing large pores to allow nucleotide leakage in bacterial walls. This disrupts the permeability of the cytoplasmic membrane
Application
  • Polymixin B sulfate targets BAC-1.2F5 murine macrophages as an LPS (lipopolysaccharide) inhibitor
  • In in vitro research, this compound is used to prevent bacterial contamination of cell cultures
  • In microbiology research, it is used as a positive and negative control during microbicidal concentration (MCC) assay
RUO – Research Use Only
Contents & Storage
Keep cold