Phosphate-Citrate Buffer (5X)
Phosphate-Citrate Buffer (5X)
Actual product may vary
Phosphate-Citrate Buffer (5X)
Used as a substrate buffer in immunoassay procedures. It is used in SDS PAGE electrophoresis.
Have Questions?
Catalog NumberQuantity
ALFJ62982.AE100 mL
Catalog number ALFJ62982.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quantity100 mL
PackagingPlastic Bottle
FormLiquid
Unit SizeEA
Contents & Storage
Ambient Temperatures