TRIS-buffered saline (TBS, 10X), with 1% Tween 20
TRIS-buffered saline (TBS, 10X), with 1% Tween 20
TRIS-buffered saline (TBS, 10X), with 1% Tween 20
TRIS-buffered saline (TBS, 10X), with 1% Tween 20
Thermo Scientific Chemicals
TRIS-buffered saline (TBS, 10X), with 1% Tween 20
Have Questions?
Catalog NumberQuantity
ALFJ62955.K32 L
Catalog number ALFJ62955.K3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 L
Request bulk or custom format
Specifications
Product TypeTBS
Quantity2 L
FormLiquid
Concentration10X
Unit SizeEA
TRIS-buffered saline (TBS, 20X), with 1% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein: protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TRIS-buffered saline (TBS, 20X), with 1% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein: protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.
Solubility
It is soluble in water.
Notes
250mM Tris, 27mM KCl, 1.37M NaCl, pH 7.4, with 1% Tween 20
RUO – Research Use Only
A Molloy.; P Laochumroonvorapong. Apoptosis, but not necrosis, of infected monocytes is coupled with killing of intracellular bacillus Calmette-Guerin. JEM. 1994, 180  ,(4), 1499-1509 .
Contents & Storage
Ambient temperatures