alpha-Terpinene, 90+%
alpha-Terpinene, 90+%
alpha-Terpinene, 90+%
alpha-Terpinene, 90+%
Thermo Scientific Chemicals
alpha-Terpinene, 90+%
CAS: 99-86-5 | C10H16 | 136.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62871.AE100 mL
Catalog number ALFJ62871.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or Materialalpha-Terpinene
CAS99-86-5
Health Hazard 1H226-H302-H304-H317-H319-H335
Health Hazard 2GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P331-P333+P313-P363-P370+P378q-P501c
View more
It can be used as a chemical fragrance essential oil and spices. It is used as a flavoring and fragrance chemical used in the personal care and cosmetic and food industries. Also used in the pharmaceutical, electronics, food processing, semi-conductor manufacturing industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used as a chemical fragrance essential oil and spices. It is used as a flavoring and fragrance chemical used in the personal care and cosmetic and food industries. Also used in the pharmaceutical, electronics, food processing, semi-conductor manufacturing industries.
Solubility
Soluble in ether, alcohol, and ethanol (100 mg/ml). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: -20°C. Store under inert gas. Air sensitive. Keep away from strong acids, strong bases.
RUO – Research Use Only
General References:
  1. H Okabe.; K Takayama.; A Ogura. Effect of limonene and related compounds on the percutaneous absorption of indomethacin. Drug design and delivery . 1989, 4 (4), 313-321.
  2. M Ondarza.; A Sanchez. Steam distillation and supercritical fluid extraction of some Mexican spices. Chromatographia. 1990, 30 (1), 16-18.