Vancomycin hydrochloride, Molecular Biology Grade
Vancomycin hydrochloride, Molecular Biology Grade
Vancomycin hydrochloride, Molecular Biology Grade
Vancomycin hydrochloride, Molecular Biology Grade
Thermo Scientific Chemicals
Vancomycin hydrochloride, Molecular Biology Grade
Vancomycin hydrochloride (vancomycin HCl, CAS # 1404-93-9) is the hydrochloride salt of the bactericidal agent, vancomycin. | CAS: 1404-93-9 | C66H76Cl3N9O24 | 1485.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFJ62790.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialVancomycin hydrochloride
TypeVancomycin Hydrochloride
Physical FormPowder
CAS1404-93-9
Health Hazard 1Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin sensitisation (category 1), Skin corrosion/irritation (category 2)
View more
Inhibits bacterial mucopeptide synthesis.Vancomycin hydrochloride is used as an antiviral and anti bacterial. binds to bacterial cell wall. It exhibits potent activity against Gram positive bacteria and is highly effective against MRSA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • Vancomycin hydrochloride is a glycopeptide antibiotic that prevents cell wall synthesis
  • Originally obtained from the bacterial strain Streptomyces Orientalis
  • Vancomycin binds to D-alanyl-D-alanine motifs in cell wall precursors and leads to cell death
  • It is active against some penicillin-resistant bacteria
Application
  • Vancomycin hydrochloride is intended for use in the research of infections and for antibiotic resistance testing
RUO – Research Use Only
General References:
  1. Yang, Z.; Liu, J.; Gao, J.; Chen, S.; Huang, G. Chitosan coated vancomycin hydrochloride liposomes: Characterizations and evaluation. Int. J. Pharm. 2015, 495 (1), 508-515.
  2. Mori, M.; Rossi, S.; Bonferoni, M. C.; Ferrari, F.; Sandri, G.; Riva, F.; Fantec, C. D.; Perotti, C.; Caramella, C. Calcium alginate particles for the combined delivery of platelet lysate and vancomycin hydrochloride in chronic skin ulcers. Int. J. Pharm. 2014, 461 (1-2), 505-513.