Sodium phosphate, 0.2M buffer soln., pH 8.0
Sodium phosphate, 0.2M buffer soln., pH 8.0
Thermo Scientific Chemicals
Sodium phosphate, 0.2M buffer soln., pH 8.0
Have Questions?
Catalog NumberQuantity
ALFJ62733.K21 L
Catalog number ALFJ62733.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium phosphate buffer soln.
Recommended StorageAmbient temperatures
Concentration0.2 M
TSCAYes
pH8.0
View more
Phosphate buffer is a multi-purpose buffer used for many molecular biology procedures. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphate buffer is a multi-purpose buffer used for many molecular biology procedures. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.
Solubility
Fully miscible in water.
Notes
Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. CR Shaw.; R Prasad. Starch gel electrophoresis of enzymes—a compilation of recipes.Biochemical genetics., 1970,4,(2), 297-320.
  2. AM Maxam.; W Gilbert. A new method for sequencing DNA. PNAS.,1977,74,(2), 560-564.