HEPES, 1.0M buffer soln., pH 7.0
HEPES, 1.0M buffer soln., pH 7.0
HEPES, 1.0M buffer soln., pH 7.0
HEPES, 1.0M buffer soln., pH 7.0
Thermo Scientific Chemicals
HEPES, 1.0M buffer soln., pH 7.0
CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number ALFJ62688.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHEPES buffer soln.
CAS7365-45-9
ColorColorless
Recommended StorageKeep cold
Concentration1.0 M
View more
HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents.  
RUO – Research Use Only
General References:
  1. Johnson, S. E.; Hudson, J. L.; Kapur, J. Synchronization of action potentials during low-magnesium-induced bursting. J. Neurophysiol. 2015, 113 (7), 2461-2470.
  2. Torelli, N. Q.; Ferreira-Júnior, J. R.; Kowaltowski, A. J.; da Cunha, F. M. RTG1-and RTG2-dependent retrograde signaling controls mitochondrial activity and stress resistance in Saccharomyces cerevisiae. Free Radical Biol. Med. 2015, 81, 30-37.