Valinomycin, 90+%
Valinomycin, 90+%
Valinomycin, 90+%
Thermo Scientific Chemicals
Valinomycin, 90+%
CAS: 2001-95-8 | C54H90N6O18 | 1111.34 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62312.MC100 mg
Catalog number ALFJ62312.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialValinomycin
TypeValinomycin
Physical FormCrystalline Powder
CAS2001-95-8
Health Hazard 1H300+H310
View more
Valinomycin, is used as a potassium ionophore shown to induce cell death. Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Valinomycin, is used as a potassium ionophore shown to induce cell death. Due to the ion-selective nature of valinomycin, it is used in ion-selective electrodes.
Solubility
Soluble in DMSO at 10mg/ml. Insoluble in w
RUO – Research Use Only
General References:
  1. Marques-Santos, L.F., et al. Cyclosporin A does not protect the disruption of the inner mitochondrial membrane potential induced by potassium ionophores in intact K562 cells. Cell Biol. Int. 2006, 30( 3), 197-204.
  2. Abdalah, R., et al. Valinomycin-induced apoptosis in Chinese hamster ovary cells. Neurosci. Lett. 2006, 405( 1-2), 68-73.