1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%
1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%
1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%
1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%
Thermo Scientific Chemicals
1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one, 98+%
Selective inhibitor of nitric oxide-sensitive guanylyl cyclase | CAS: 41443-28-1 | C9H5N3O2 | 187.158 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ62250.MA10 mg
Catalog number ALFJ62250.MA
Price (MYR)
837.00
EA
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
837.00
EA
Add to cart
Specifications
Chemical Name or Material1H-[1,2,4]Oxadiazolo[4,3-a]-quinoxalin-1-one
CAS41443-28-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Selective inhibitor of nitric oxide-sensitive guanylyl cyclase