Paraformaldehyde fixative solution
Paraformaldehyde fixative solution
Thermo Scientific Chemicals
Paraformaldehyde fixative solution
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number ALFJ61984.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialParaformaldehyde fixative solution
Physical FormLiquid
Health Hazard 1H317-H318-H351
Health Hazard 2GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
View more
Paraformaldehyde fixative solution finds application in proteins, peptides and the enzymes of low molecular weight.