Streptozotocin, 97+%
Streptozotocin, 97+%
Streptozotocin, 97+%
Streptozotocin, 97+%
Thermo Scientific Chemicals
Streptozotocin, 97+%
Streptozotocin (streptozocin, CAS # 18883-66-4) is an N-nitrosourea antibiotic produced by Streptomyces achromogenes. | CAS: 18883-66-4 | C8H15N3O7 | 265.222 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Catalog number ALFJ61601.ME
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialStreptozotocin
TypeStreptozotocin
Physical FormPowder
CAS18883-66-4
Health Hazard 1H350
View more
Streptozotocin is used to induce Type 1 diabetes for medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Streptozotocin acts as a bactericidal by alkylating DNA and inhibiting DNA synthesis
• Streptozotocin enters cells through glucose transporter 2
Application
• Streptozotocin induces diabetes in experimental animals and can cause acute liver injury
• It is also useful for research on neuroendocrine tumors and carcinoma cells
RUO – Research Use Only
General References:
  1. Duraisamy, A.J.; Mohammad, G.; Kowluru, R.A. Mitochondrial fusion and maintenance of mitochondrial homeostasis in diabetic retinopathy. Biochim Biophys Acta Mol Basis Dis. 2019, 1865(6), 1617-1626.
  2. Zhu, Y.; Han, X.Q.; Sun, X.J.; Yang, R.; Ma, W.Q.; Liu, N.F. Lactate accelerates vascular calcification through NR4A1-regulated mitochondrial fission and BNIP3-related mitophagy. Apoptosis. 2020, (5-6), 321-340.