D-Amygdalin, 98%
D-Amygdalin, 98%
D-Amygdalin, 98%
Thermo Scientific Chemicals
D-Amygdalin, 98%
A benzylic glycoside that has been used as an antineoplastic agent | CAS: 29883-15-6 | C20H27NO11 | 457.43 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61472.031 g
Catalog number ALFJ61472.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Amygdalin
CAS29883-15-6
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
A benzylic glycoside that has been used as an antineoplastic agent