Potassium phosphate, 0.5M buffer soln., pH 7.4
Potassium phosphate, 0.5M buffer soln., pH 7.4
Potassium phosphate, 0.5M buffer soln., pH 7.4
Potassium phosphate, 0.5M buffer soln., pH 7.4
Thermo Scientific Chemicals
Potassium phosphate, 0.5M buffer soln., pH 7.4
CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ61413.AP500 mL
Catalog number ALFJ61413.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium phosphate
CAS7778-77-0
Recommended StorageAmbient temperatures
Concentration0.5 M
pH7.4
View more
Potassium phosphate is used as a buffer for the determination of pH. It is also used in pharmaceutical production, urinary acidifier, paper processing, nutrient solutions and yeast foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium phosphate is used as a buffer for the determination of pH. It is also used in pharmaceutical production, urinary acidifier, paper processing, nutrient solutions and yeast foods.
Solubility
Miscible with water. Immiscible with alcohol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Pankratov, D.; Sundberg, R.; Sotres, J.; Maximov, I.; Graczyk, M.; Suyatin, D. B.; González-Arribas, E.; Lipkin, A.; Montelius, L.; Shleev, S. Transparent and flexible, nanostructured and mediatorless glucose/oxygen enzymatic fuel cells. J. Power Sources 2015, 294, 501-506.
  2. Vu, T. B. N.; Do, T. T.; Le, T. H.; Nguyen, T. T.; Nguyen, S. L. T. Enhance production of recombinant lumbrokinase by optimizing gene codon usage for expression in Pichia pastoris and its properties. J. Biotech Res. 2015, 6, 96-106.