Carbonate-buffered saline (5X), pH 9.0
Carbonate-buffered saline (5X), pH 9.0
Actual product may vary
Carbonate-buffered saline (5X), pH 9.0
Carbonate-buffered saline (5X), pH 9.0
Carbonate-buffered saline (5X), pH 9.0
Have Questions?
Catalog NumberQuantity
ALFJ61396.AK250 mL
Catalog number ALFJ61396.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Concentration5X
pH Range9.0
FormatLiquid
Quantity250 mL
Storage RequirementsAmbient temperatures
TypeCarbonate-buffered saline
Unit SizeEA