TRIS, 0.5M buffer soln., pH 7.0, Thermo Scientific™
TRIS, 0.5M buffer soln., pH 7.0, Thermo Scientific™
TRIS, 0.5M buffer soln., pH 7.0, Thermo Scientific™
TRIS, 0.5M buffer soln., pH 7.0, Thermo Scientific™
TRIS, 0.5M buffer soln., pH 7.0, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFJ61213.K21 L
Catalog number ALFJ61213.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialTRIS buffer soln.
CAS77-86-1
Recommended StorageAmbient temperatures
Concentration0.5 M
TSCAYes
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.