Sodium phosphate, 0.5M buffer soln., pH 8.5
Sodium phosphate, 0.5M buffer soln., pH 8.5
Thermo Scientific Chemicals
Sodium phosphate, 0.5M buffer soln., pH 8.5
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number ALFJ61151.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium phosphate buffer soln.
Recommended StorageAmbient temperatures
Concentration0.5 M
TSCAYes
pH8.5