Tropicamide, 99+%
Tropicamide, 99+%
Tropicamide, 99+%
Thermo Scientific Chemicals
Tropicamide, 99+%
CAS: 1508-75-4 | C17H20N2O2
Have Questions?
Catalog NumberQuantity
ALFJ61132.031 g
Catalog number ALFJ61132.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Quantity1 g
DescriptionWhite
For Use With (Application)A muscarinic antagonist
Recommended StorageKeep cold
Assay≥99%
View more
A muscarinic antagonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C