Agarose, high melting temperature, medium resolution
Agarose, high melting temperature, medium resolution
Agarose, high melting temperature, medium resolution
Agarose, high melting temperature, medium resolution
Thermo Scientific Chemicals
Agarose, high melting temperature, medium resolution
For standard DNA applications | CAS: 9012-36-6
Have Questions?
Catalog NumberQuantity
ALFJ61123.36500 g
Catalog number ALFJ61123.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAgarose
Name Notehigh melting temperature, medium resolution
Physical FormPowder
CAS9012-36-6
Recommended StorageAmbient temperatures
View more
For standard DNA applications.Agarose, High Melting Temperature, Medium Resolution is used for standard DNA separations. It is involved in the purification of protein by using column-based preparative scale separation as in gel filtration chromatography and ion exchange chromatography
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
For standard DNA applications.Agarose, High Melting Temperature, Medium Resolution is used for standard DNA separations. It is involved in the purification of protein by using column-based preparative scale separation as in gel filtration chromatography and ion exchange chromatography
Solubility
Soluble in water.
Notes
Melting temp: ∼88°.Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Hernández, C.; Alvarez, C.; González, C.; Ayala, M. S.; León, C. M.; Ramírez, J. D. Identification of Six New World Leishmania species through the implementation of a High-Resolution Melting (HRM) genotyping assay. Parasites Vectors 2014, 7 (1), 1-7.
  2. Alnuaimi, A. D.; Wiesenfeld, D.; O'Brien-Simpson, N. M.; Reynolds, E. C.; Peng, B.; McCullough, M. J. The development and validation of a rapid genetic method for species identification and genotyping of medically important fungal pathogens using high-resolution melting curve analysis. Mol Oral Microbiol 2014, 29 (3), 117-130.