Acetate Buffer (1 M, pH 5.5)
Acetate Buffer (1 M, pH 5.5)
Acetate Buffer (1 M, pH 5.5)
Acetate Buffer (1 M, pH 5.5)
Thermo Scientific Chemicals
Acetate Buffer (1 M, pH 5.5)
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500mL
250mL
Catalog number ALFJ61033.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500mL
Request bulk or custom format
Specifications
CAS127-09-3
Concentration1M buffer soln.
Molecular FormulaC2H3NaO2
MDL NumberMFCD00012459
InChI KeyVMHLLURERBWHNL-UHFFFAOYSA-M
View more
Acetate, 1M buffer soln., pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acetate, 1M buffer soln., pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.
Notes
Contains sodium acetate and glacial acetic acid, pH 5.5
RUO – Research Use Only
General References:
  1. Tong Ihn Lee et. al. Chromatin immunoprecipitation and microarray-based analysis of protein location. Nature Protocols. 2006, 1,(2), 729-748.
  2. Jaspreet Kaur et. al. In silico screening, structure-activity relationship, and biologic evaluation of selective pteridine reductase inhibitors targeting visceral leishmaniasis. Antimicrobial Agents and Chemotherapy. 2011, 55,(2), 659-666.