Borate-buffered saline (5X)
Borate-buffered saline (5X)
Actual product may vary
Borate-buffered saline (5X)
Borate-buffered saline (5X)
Borate-buffered saline (5X)
Have Questions?
Catalog NumberQuantity
ALFJ60979.AK250 mL
Catalog number ALFJ60979.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Borate buffered saline has been used to determine whether heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis can be used to treat canine flea allergy. Borate buffered saline has also been used to investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. Borate buffered saline has been used to determine the effects of packaging solution osmolality and buffering agent on soft contact lens parameters. Borate buffer solutions is used in protein modification procedures requiring amine-free buffer at alkaline pH.
Solubility
Soluble in water 0.1 M at 20°C.
Notes
125mM sodium borate, 750mM sodium chloride, pH 7.0
General References:
  1. Susan M. Kaska,; S. Sarangapani,; José Giner. Oxygen Reduction on Platinum in Borate-Buffered Saline Solutions. J. Electrochem. Soc. 1989, 136 (1), 75-83.
  2. Huang, X.Y.,; et al. Zhonghua,; Gan Zang Bing Za Zhi .Effects of MIM-B gene on invasive and metastatic potentials of human hepatocellular carcinoma MHCC97H cells . 2010 , 18915-9.
Specifications
Product TypeBorate-buffered Saline
Quantity250 mL
FormLiquid
Concentration5X
Molecular FormulaB4Na2O7
MDL NumberMFCD00081185
InChI KeyNXSVMNYRVBQCMO-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
IUPAC Namedisodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular Weight (g/mol)201.21
Unit SizeEA
Contents & Storage
Ambient temperatures