(+/-)-Miconazole nitrate, 98+%
(+/-)-Miconazole nitrate, 98+%
(+/-)-Miconazole nitrate, 98+%
(+/-)-Miconazole nitrate, 98+%
Thermo Scientific Chemicals
(+/-)-Miconazole nitrate, 98+%
Inhibits cytochrome P450-dependent 14a-demethylase | CAS: 22832-87-7 | C18H15Cl4N3O4 | 479.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60796.065 g
Catalog number ALFJ60796.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-Miconazole nitrate
CAS22832-87-7
Health Hazard 1H302-H317
Health Hazard 2GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
View more
Inhibits cytochrome P450-dependent 14a-demethylase