Resveratrol, 98%
Resveratrol, 98%
Resveratrol, 98%
Thermo Scientific Chemicals
Resveratrol, 98%
CAS: 501-36-0 | C14H12O3 | 228.247 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60790.ME500 mg
Catalog number ALFJ60790.ME
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialResveratrol
CAS501-36-0
Health Hazard 1H302-H315-H317-H318-H335
Health Hazard 2GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
View more
It is used as a natural phenolic antioxidant that is a potent activator of SIRT1 and inhibitor of Cox-1. Resveratrol has also been used extensively in stem cell studies to enhance self-renewal and multipotency of mesencymal stem cells (MSC’s) in vitro. Resveratrol acts as a NF-κB inhibitor to protect MSC-derived chondrocytes in vitro. Resveratrol is used to treat adipose-derived mesenchymal stem cells (ADMSCs) resulted in increased alkaline phosphatase activity and osteocalcin levels; thus enriching the MSC and osteoprogenitor cell populations in cultured ADMSCs in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a natural phenolic antioxidant that is a potent activator of SIRT1 and inhibitor of Cox-1. Resveratrol has also been used extensively in stem cell studies to enhance self-renewal and multipotency of mesencymal stem cel
RUO – Research Use Only
General References:
  1. Kimura et al. Effects of stilbenes on arachidonate metabolism in leukocytes. Biochim.Biophys.Acta . 1985, 834 ,(2), 275-8.
  2. Chun et al . Resveratrol is a selective human cytochrome P450 1A1 inhibitor. Biochem.Biophys.Res.Commun.1999, 262 ,(1), 20-4.