NP-40 lysis buffer
Thermo Scientific Chemicals
NP-40 lysis buffer
Have Questions?
Catalog NumberQuantity
ALFJ60766.AP500 mL
Catalog number ALFJ60766.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
ColorColorless
FormatLiquid
Quantity500 mL
Unit SizeEA
NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.
Solubility
It is soluble in water.
Notes
This buffer contains: 50mM Tris-HCl (pH 7.4), 150mM NaCl, 1%NP-40 and 5mM EDTA. To use, incubate cells in 5 volumes of buffer on ice for 10-30 minutes.
RUO – Research Use Only
General References:
  1. M Herrmann.; HM Lorenz.; R Voll.; M Grünke. A rapid and simple method for the isolation of apoptotic DNA fragments. Nucleic Acids Res. 1994, 22,(24), 5506-5507.
  2. K Guo.; J Wang.; V Andres.; RC Smith. MyoD-induced expression of p21 inhibits cyclin-dependent kinase activity upon myocyte terminal differentiation. Mol. Cell. Biol. 1995, 15,(7), 3823-3829.
Contents & Storage
Keep cold