Acrylamide/Bisacrylamide 19:1, powder
Acrylamide/Bisacrylamide 19:1, powder
Acrylamide/Bisacrylamide 19:1, powder
Acrylamide/Bisacrylamide 19:1, powder
Thermo Scientific Chemicals
Acrylamide/Bisacrylamide 19:1, powder
Premixed powder for preparing protein PAGE gels
Have Questions?
Catalog NumberQuantity
ALFJ60486.22100 g
Catalog number ALFJ60486.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAcrylamide/Bisacrylamide 19:1
Physical FormPowder
Health Hazard 1H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372
Health Hazard 2GHS H Statement
H301-H340-H350-H372-H361-H312-H332-H315-H319-H317
Toxic if swallowed.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
View more
Premixed powder for preparing protein PAGE gels