Sodium cacodylate, 0.1M buffer soln., pH 6.5
Sodium cacodylate, 0.1M buffer soln., pH 6.5
Sodium cacodylate, 0.1M buffer soln., pH 6.5
Sodium cacodylate, 0.1M buffer soln., pH 6.5
Thermo Scientific Chemicals
Sodium cacodylate, 0.1M buffer soln., pH 6.5
CAS: 6131-99-3214.024 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ60367.AD50 mL
Catalog number ALFJ60367.AD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Specifications
Concentration0.1M buffer soln.
Molecular Weight (g/mol)214.024
pH6.5
Quantity50 mL
CAS6131-99-3
View more
Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.
Solubility
Soluble in water (∼100 g/L) at 20°C, ethanol (400 mg/mL), and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only
General References:
  1. WG Miller.; MA Cusanovich. Electron transport by C-type cytochromes. Biophysics of structure and mechanism. 1975, 1,(2), 97-111.
  2. RW Van Dyke. Acidification of rat liver lysosomes: quantitation and comparison with endosomes. American Journal of Physiology-Cell. 1993, 265,(4), 901-917.
Contents & Storage
Keep cold