BioAcryl-P (30%, 19:1)
BioAcryl-P (30%, 19:1)
BioAcryl-P (30%, 19:1)
BioAcryl-P (30%, 19:1)
Thermo Scientific Chemicals
BioAcryl-P (30%, 19:1)
Premixed solution for preparing protein PAGE gels
Have Questions?
Catalog NumberQuantity
ALFJ60126.K21 L
Catalog number ALFJ60126.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialAcrylamide/Bisacrylamide 19:1
Physical FormLiquid
Health Hazard 1H302+H332-H315-H317-H319-H340-H350-H361f-H372
Health Hazard 2GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
View more
Premixed solution for preparing protein PAGE gels