Alcian Blue 8GX
Alcian Blue 8GX
Alcian Blue 8GX
Alcian Blue 8GX
Thermo Scientific Chemicals
Alcian Blue 8GX
CAS: 33864-99-2 | C56H68Cl4CuN16S4 | 1298.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Catalog number ALFJ60122.22
Price (MYR)
3,782.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
3,782.00
EA
Add to cart
Specifications
Chemical Name or MaterialAlcian Blue 8GX
Melting Point148°C
CAS33864-99-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Alcian Blue 8GX is widely utilized as a heteroglycan stain for acidic, neutral, sulfated and phosphate polysaccharides and glycosaminoglycans in tissues. It finds application in both light and electron microscopy. It serves as an adhesive to stick glycol methacrylate sections to glass slides. Due to its large aromatic surface, it is used as a coating agent and modifies the property on the surfaces. It acts as a gelling agent for lubricating fluids and is also used in the modification of carbon paste electrode which promotes redox behavior and stability of electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Alcian Blue 8GX is widely utilized as a heteroglycan stain for acidic, neutral, sulfated and phosphate polysaccharides and glycosaminoglycans in tissues. It finds application in both light and electron microscopy. It serves as an adhesive to stick glycol methacrylate sections to glass slides. Due to its large aromatic surface, it is used as a coating agent and modifies the property on the surfaces. It acts as a gelling agent for lubricating fluids and is also used in the modification of carbon paste electrode which promotes redox behavior and stability of electrode.
Solubility
Soluble in water, ethanol, ethylene glycol and 2-ethoxyethanol. Insoluble in xylene.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Kameyama , A.; Dong, W.; Matsuno, Y. Succinylation-Alcian Blue Staining of Mucins on Polyvinylidene Difluoride Membranes. Methods Mol. Biol. 2015, 1314, 325-331.
  2. Candiano, G.; Santucci, L.; Petretto, A.; Inglese, E.; Bruschi, M.; Ghiggeri, G. E.; Boschetti, E.; Righetti, G. Widening and Diversifying the Proteome Capture by Combinatorial Peptide Ligand Libraries via Alcian Blue Dye Binding. Anal. Chem. 2015, 87 (9), 4814-4820