Enrofloxacin
Enrofloxacin
Enrofloxacin
Thermo Scientific Chemicals
Enrofloxacin
A plant fungicide and antibacterial agent | CAS: 93106-60-6 | C19H22FN3O3 | 359.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFJ60023.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEnrofloxacin
TypeEnrofloxacin
Physical FormPowder
CAS93106-60-6
Melting Point221°C to 226°C
View more
Enrofloxacin is a broad spectrum antibiotic bactericidal agent used in veterinary medicine to treat animals afflicted with certain bacterial infections.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Enrofloxacin is a broad spectrum antibiotic bactericidal agent used in veterinary medicine to treat animals afflicted with certain bacterial infections.
Solubility
Soluble in chloroform. Slightly soluble in water. Also soluble in dilute KOH
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Walker RD; Stein GE; Hauptman JG; MacDonald KH. Pharmacokinetic evaluation of enrofloxacin administered orally to healthy dogs. American Journal of Veterinary Research.199253(12), 2315-2319.
  2. Mengozzi G; Intorre L; Bertini S; Soldani G. Pharmacokinetics of enrofloxacin and its metabolite ciprofloxacin after intravenous and intramuscular administrations in sheep. American Journal of Veterinary Research.199657(7), 1040-1043.