Glycine, ≥99%, Molecular Biology Grade, Ultrapure
Glycine, ≥99%, Molecular Biology Grade, Ultrapure
Glycine, ≥99%, Molecular Biology Grade, Ultrapure
Glycine, ≥99%, Molecular Biology Grade, Ultrapure
Thermo Scientific Chemicals
Glycine, ≥99%, Molecular Biology Grade, Ultrapure
CAS: 56-40-6 | C2H5NO2 | 75.067 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALFJ16407.A75 kgPlastic Bottle
Catalog number ALFJ16407.A7
Price (MYR)
2,693.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
2,693.00
EA
Add to cart
Specifications
Chemical Name or MaterialGlycine
Melting Point∼245°C (decomposition)
CAS56-40-6
Recommended StorageAmbient temperatures
RTECS NumberMB7600000