L-Lysine ethyl ester diisocyanate, 97%
L-Lysine ethyl ester diisocyanate, 97%
L-Lysine ethyl ester diisocyanate, 97%
L-Lysine ethyl ester diisocyanate, 97%
Thermo Scientific Chemicals
L-Lysine ethyl ester diisocyanate, 97%
CAS: 45172-15-4 | C10H14N2O4 | 226.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH64384.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Lysine ethyl ester diisocyanate
CAS45172-15-4
Health Hazard 1H315-H317-H319-H331-H334-H335
Health Hazard 2GHS H Statement
H331-H334-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P311-P333+P313-P362-P501c
View more
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Sparingly soluble in water (0.19 g/L).
Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Jian Han; Bing Chen; Lin Ye; Ai-ying Zhang; Jian Zhang; Zeng-guo Feng. Synthesis and characterization of biodegradable polyurethane based on poly(ε-caprolactone) and L-lysine ethyl ester diisocyanate. Frontiers of Materials Science in China. 20093 (1), 25-32.
  2. R. F. Storey; J. S. Wiggins; and A. D. Puckett. Hydrolyzable poly(ester-urethane) networks from L-lysine diisocyanate and D,L-lactide/ϵ-caprolactone homo- and copolyester triols. Journal of Polymer Science Part A: Polymer Chemistry. 199432 (12), 2345-2363.