1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals
1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals
1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals
1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFH64116.1425 g
Catalog number ALFH64116.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1-(Bromomethyl)naphthalene
CAS3163-27-7
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
It is employed as a intermediate for organic synthesis and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for organic synthesis and pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Linda J. Johnston; J. C. Scaiano. Generation, spectroscopy, and reactivity of excited 1-naphthylmethyl radicals. Org. Lett. 1985107(22), 6368-6372.
  2. Takashi Ooi; Yuichiro Arimura; Yukihiro Hiraiwa; Lin Ming Yuan; Taichi Kano; Toru Inoue; Jun Matsumoto; Keiji Maruoka. Highly enantioselective monoalkylation of p-chlorobenzaldehyde imine of glycine tert-butyl ester under mild phase-transfer conditions. Tetrahedron: Asymmetry. 200617(4), 603-606.