2-Bromo-4'-(trifluoromethoxy)acetophenone, 97%
2-Bromo-4'-(trifluoromethoxy)acetophenone, 97%
2-Bromo-4'-(trifluoromethoxy)acetophenone, 97%
2-Bromo-4'-(trifluoromethoxy)acetophenone, 97%
Thermo Scientific Chemicals
2-Bromo-4'-(trifluoromethoxy)acetophenone, 97%
CAS: 103962-10-3 | C9H6BrF3O2 | 283.044 g/mol
Have Questions?
Catalog NumberQuantity
ALFH64105.031 g
Catalog number ALFH64105.03
Price (MYR)
519.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
519.00
EA
Add to cart
Specifications
Chemical Name or Material2-Bromo-4'-(trifluoromethoxy)acetophenone
CAS103962-10-3
Health Hazard 1H314
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c