N-Benzoyl-L-tyrosine ethyl ester, 98%
N-Benzoyl-L-tyrosine ethyl ester, 98%
N-Benzoyl-L-tyrosine ethyl ester, 98%
N-Benzoyl-L-tyrosine ethyl ester, 98%
Thermo Scientific Chemicals
N-Benzoyl-L-tyrosine ethyl ester, 98%
CAS: 3483-82-7 | C18H19NO4 | 313.35 g/mol
Have Questions?
Catalog NumberQuantity
ALFH63434.065 g
Catalog number ALFH63434.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Benzoyl-L-tyrosine ethyl ester
Melting Point118°C to 121°C
CAS3483-82-7
Storage Note 1-30° to -10°C
Recommended StorageStore at -20°C