10-Hydroxydecanoic acid, 96%
10-Hydroxydecanoic acid, 96%
10-Hydroxydecanoic acid, 96%
10-Hydroxydecanoic acid, 96%
Thermo Scientific Chemicals
10-Hydroxydecanoic acid, 96%
CAS: 1679-53-4 | C10H20O3 | 188.27 g/mol
Have Questions?
Catalog NumberQuantity
ALFH60743.031 g
Catalog number ALFH60743.03
Price (MYR)
629.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
629.00
EA
Add to cart
Specifications
Chemical Name or Material10-Hydroxydecanoic acid
CAS1679-53-4
Melting Point75°C to 77°C
Recommended StorageAmbient temperatures
EINECS Number216-848-1