1-Ethyl-3-methylimidazolium iodide, 97%
1-Ethyl-3-methylimidazolium iodide, 97%
1-Ethyl-3-methylimidazolium iodide, 97%
1-Ethyl-3-methylimidazolium iodide, 97%
Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium iodide, 97%
CAS: 35935-34-3 | C6H11IN2 | 238.07 g/mol
Have Questions?
Catalog NumberQuantity
ALFH59404.065 g
Catalog number ALFH59404.06
Price (MYR)
289.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
289.00
EA
Add to cart
Specifications
Chemical Name or Material1-Ethyl-3-methylimidazolium iodide
CAS35935-34-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is used as OLED materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as OLED materials.
Solubility
Soluble in water, , dichloromethane, ethanol, acetonitrile.
Notes
Moisture Sensitive. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Annegret Stark.; Bonnie L. MacLean.; Robert D. Singer. 1-Ethyl-3-methylimidazolium halogenoaluminate ionic liquids as solvents for Friedel-Crafts acylation reactions of ferrocene. J. Chem. Soc., Dalton Trans. 1999, (1),63-66.
  2. Laine Green.; Ivan Hemeon.; Robert D Singer. 1-Ethyl-3-methylimidazolium halogenoaluminate ionic liquids as reaction media for the acylative cleavage of ethers.Tetrahedron Lett. 2000, 41 (9), 1343-1346.